Learn More
Techne™ Stuart™ Skirted 96-well PCR Plates
These 96-well PCR plates are skirted to reduce plate distortion and provide stability.
2455.00 SEK - 3445.00 SEK
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
No. of Wells | 96 |
No. per Pack | 25 |
Profile | Low |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
13048523
|
Techne™
16210-30 |
Black |
3445.00 SEK
Pack of 25 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
13038523
|
Techne™
10125-26 |
Clear |
3395.00 SEK
Pack of 25 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
13058523
|
Techne™
16210-31 |
White |
2455.00 SEK
Pack of 25 |
Estimated Shipment: 05-11-2025
Log in to see stock. |
|||||
Description
- Fully skirted to reduce distortion and promote stability
- Available in Black, Clear and White
Specifications
Standard | |
96 | |
Low |
8 x 12 | |
25 | |
25 Pk. |