Learn More
Cole-Parmer™ Stuart™ Accessories Set
Improve and expand the performance of you vortex mixer
Brand: Cole-Parmer™ 04729-07
1104.07 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with the variable-speed vortexer.
Kit contains:
- 1x plastic cradle for standard microtitre plate
- 1x circular foam insert for beakers and flasks (up to 500 mL capacity)
- 1x rectangular foam insert (holds 8 x 0.2 mL, 8 x 0.5 mL and 16 x 1.5 mL microcentrifuge tubes)
- 1x solid rectangular foam insert (blank for custom holes)
Please note: The microtitre plate holder SA8/1 should not be used at speeds above 800 rpm.
Specifications
SA8 Vortex Mixer |