Learn More
STOML1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12862-1-AP-20UL
This item is not returnable.
View return policy
Specifications
STOML1 | |
Polyclonal | |
Unconjugated | |
STOML1 | |
EPB72 like protein 1, hUNC 24, Protein unc 24 homolog, SLP 1, SLP1, stomatin (EPB72) like 1, Stomatin like protein 1, Stomatin related protein, STOML1, STORP, UNC24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
300748, 69106, 9399 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CI66, Q9UBI4 | |
STOML1 | |
STOML1 Fusion Protein Ag3978 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |