Learn More
Sampling Systems™ SteriWare™ Disposable Jugs
Comfortably hold this jug due to its rounded handle. Sampling Systems™ SteriWare™ Disposable Jugs have a wide base, which makes them very stable.
815.00 SEK - 2040.00 SEK
Specifications
Capacity (Metric) | 2000 mL |
---|---|
Includes | Individually Bagged |
Material | HDPE |
Disposable | Single-use |
Graduated | No |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15306785
![]() |
Sampling Systems™
8049H-2000S |
16 Per Box |
2040.00 SEK
Pack of 16 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
15340024
![]() |
Sampling Systems™
8049H-2000 |
20 Per Box |
815.00 SEK
Pack of 20 |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No Cross Contamination
- No Cleaning required
Specifications
2000 mL | |
HDPE | |
No | |
129 mm |
Individually Bagged | |
Single-use | |
152 mm |