Learn More
Gammex™ Sterile Latex Surgical Gloves
Gammex™ Sterile Latex Surgical Gloves are advanced powder-free latex gloves for general surgery with a micro-rough outer surface for enhanced grip.
Brand: Gammex™ AJ330048060
This item is not returnable.
View return policy
Description
- Advanced powder-free latex surgical gloves
- Optimal combination of safety and comfort
- Provides a secure barrier without compromising sensitivity
- Donning is made easy, even with damp hands, thanks to the water-based polyurethane inner coating
- Micro-rough outer surface for enhanced grip
- Very low latex protein and chemical accelerator residue content safe on skin
- For laboratory research purposes only
Specifications
AS/NZS 4179, ASTM F739, EN 374-1/2/3, EN 420, EN 455 1-4, ISO 10282, JIS T9107 | |
Single-use | |
No | |
Powder Free | |
Sterile |
Natural | |
Ambidextrous | |
Latex | |
6 | |
Straight |