Learn More
Sigma Laborzentrifugen™ Steel Centrifuge Tables
Use these centrifuge tables for a wide variety of applications. Sigma™ Steel Centrifuge Tables are made of sheet steel with two doors and space for accessories.
1470.00 €
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
10195832
|
Sigma Laborzentrifugen™
17920 |
Sigma 2-16K, Sigma 2-16KC, Sigma 2-16KCH | 550mm W x 600mm D x 497mm H |
1470.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10811351
|
Sigma Laborzentrifugen™
17927 |
Sigma 3-16, Sigma 3-18 | 490mm W x 684mm D x 515mm H |
1470.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- With 2 doors
- Manufactured from varnished sheet steel
- With space for accessories
- Movable on lockable castors
Specifications
Centrifuge Table | |
Steel |