Learn More
Fisherbrand™ Stainless Steel Test Sieve Stacks
Use these sieve stacks for research and development, quality control of raw materials, interim and finished products as well as in production monitoring. Fisherbrand™ Stainless Steel Test Sieve Stacks are designed for use with AS 200 control vibratory sieve shakers.
Brand:  Fisherbrand™ 60.131.000999
Product Code. 10670452
  • 14175.20 SEK / Each
Only null left
Estimated Shipment: 15-07-2025
Description
  • Sieve stack consisting of 8 test sieves (ISO 3310-1)
  • 200mm diameter, 50mm height (45μm, 63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm)
  • Includes collecting pan
Specifications
Specifications
8 Sieves
Test Sieve Stack
Certificate of compliance with ISO 3310/1 or NFX 11-504, 8 Sieves (45 μm, 63 μm, 125 μm, 250 μm, 500 μm, 1 mm, 2 mm, 4 mm) and collecting pan
For separation, fractioning, particle size determination
Stainless Steel
45 μm, 63 μm, 125 μm, 250 μm, 500 μm, 1 mm, 2 mm, 4 mm
ISO 3310 and ASTM E 11
50 mm
200 mm