Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN30, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SN30B02070
Product Code. 17518595
  • 82.00€ / Each
Estimated Shipment: 24-06-2025
Explore more special offers
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SN30 Sterilizer
Drip Tray
Rim: 15 mm