Learn More
SPEX™ Stainless Steel Grinding Ball
SPEX™ Stainless Steel Grinding Ball is a replacement ball for use with 3114 Stainless Steel Grinding Vial Set.
Brand:  SPEX™ 3114SB
223.37 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15390522
  • 240.00 SEK / Pack of 4
Only null left
Estimated Shipment: 20-08-2025
Description
  • Durable and tough grinding ball suitable for general purpose grinding
  • High durability and efficiency with moderate resistance to abrasion
  • Resists chemical attack
  • Typical composition: Major Elements – Fe, Cr; Minor Elements – Ni, Mn, S, Si
  • Hardness: Mohs 5-5 1/2; Rockwell – C 55-60
Specifications
Specifications
6.35 mm
3114 Stainless Steel Grinding Vial Set
Stainless Steel
4 Balls