Learn More
GFL Gesellschaft Fuer Labortec™ Stainless Steel Fume Hoods for Waterbath
Use these baths for a varied range of applications. GFL Gesellschaft fuer Labortec™ Stainless Steel Fume Hoods for Waterbaths are ideal for gentle steaming work with Erlenmeyer flasks, glasses, etc.
11970.00 SEK - 14245.00 SEK
Specifications
Ambient Temperature | 5°C to 99.9°C |
---|---|
Controller Type | Tension Thermostat |
Description | Waterbath |
For Use With (Equipment) | Fume Hoods |
Temperature Range | Ambient +5° to 99.9°C |
Description
- Bath, removable recessed lid with holes and perforated floor above the heating element are made of stainless steel
- Adjustable water level regulator at the right-hand side of the unit is part of the standard scope of supply
- Openings in the lid are covered by sets of rings made of heat-resistant plastic material
- Temperature range from approx. 5°C above ambient to boiling point, temperature regulation through
- tension thermostat
- Four non-slip plastic feet ensure great stability
- Heating element protected by over-temperature cut-out
Specifications
5°C to 99.9°C | |
Waterbath | |
Ambient +5° to 99.9°C | |
Stainless Steel | |
100 mm | |
230 V | |
50/60 Hz |
Tension Thermostat | |
Fume Hoods | |
Gentle steaming work with Erlenmeyer flasks, beakers, etc. | |
155 mm | |
300 mm | |
1.5 kW |