Learn More
Julabo™ Stainless Steel 1-Adaptor
1340.00 SEK - 1540.00 SEK
Specifications
For Use With (Equipment) | Suitable for CF30, CF31, CF40, CF41 Cryo-Compact Circulator |
---|---|
Type | 1-Adapter |
Material | Stainless Steel |
Product Code | Brand | Connection Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Connection Type | Price | Quantity & Availability | |||||
10752604
|
Julabo™
8891008 |
M16 x 1 Male to 1/2 in. BSP Female |
1340.00 SEK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
|||||
11910288
|
Julabo™
8891009 |
M16 x 1 Male to 3/4 in. BSP Female |
1540.00 SEK
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
|||||
Specifications
Suitable for CF30, CF31, CF40, CF41 Cryo-Compact Circulator | |
1-Adapter | |
Stainless Steel |