Learn More
SRF Mouse anti-Human, CoraLite 488, Clone: 1G8F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66742-100UL
Description
The SRF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SRF. This antibody recognizes human antigen. The SRF antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
SRF | |
Monoclonal | |
2000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
1G8F4 | |
CoraLite 488 | |
100 μL | |
6722 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.