Learn More
Buerkle™ Sporty Style Safety Goggles
Shop All Bürkle Products
Click to view available options
:
Color: Blue and orange
Description
- Ultra-light protective goggles
- Sporty design
- Maximum wearing comfort
- According to DIN/EN standard
Specifications
Specifications
Certifications/Compliance | DIN/EN standards |
Lens Tint | Clear |
Product Type | Casual style safety goggles |
For Use With (Application) | Eye Protection |
Frame Color | Blue and orange |
No. per Pack | 1 Ea. |
UV Protection | None |