Learn More
JP Selecta™ Spatula with Spoon and Flat Ends
Suitable for general laboratory purpose. JP Selecta™ Spatula with Spoon and Flat Ends is made of AISI 304 stainless steel.
10.00€ - 19.00€
Specifications
Product Type | Flat End/Spoon Ended Spatula |
---|---|
Material | Stainless Steel (304) |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12037554
|
JP Selecta™
1096120 |
120 mm |
10.00€
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
|||||
12047554
|
JP Selecta™
1096150 |
150 mm |
19.00€
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
|||||
Description
- Made of AISI 304 stainless steel
Specifications
Flat End/Spoon Ended Spatula | |
Stainless Steel (304) |