Learn More
Sodium phosphate, 0.2M buffer soln., pH 9.0, Thermo Scientific™
768.00 SEK - 1060.00 SEK
Specifications
Chemical Name or Material | Sodium phosphate buffer soln. |
---|---|
Concentration | 0.2 M |
pH | 9.0 |
Physical Form | Liquid |
MDL Number | MFCD00146206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15443309
|
Thermo Scientific Alfa Aesar
J60565.AP |
500 mL |
768.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453309
|
Thermo Scientific Alfa Aesar
J60565.K2 |
1 L |
1060.00 SEK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sodium phosphate buffer soln. | |
9.0 | |
MFCD00146206 |
0.2 M | |
Liquid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures