Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 7.0
1165.00 SEK - 1485.00 SEK
Specifications
Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.2 M
pH 7.0
Physical Form Liquid
MDL Number MFCD00146206
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15425709
View Documents
Thermo Scientific Chemicals
J63482.AP
500 mL
1165.00 SEK
500mL
15435709
View Documents
Thermo Scientific Chemicals
J63482.K2
1 L
1485.00 SEK
1 L
Description
Description
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphate is widely used as a reagent with high buffering capacity for molecular biology, biochemistry, and chromatography
Solubility
Fully miscible.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Sodium phosphate buffer soln.
7.0
MFCD00146206
0.2 M
Liquid
Fully miscible.
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures