Learn More
Sodium nitrite, BAKER ANALYZED™ ACS, J.T. Baker™
Product Code. 10784581
Click to view available options
Quantity:
100 g
1 kg
Exceeds ACS Specifications. Meets Reagent Specifications for testing USP/NF monographs
Chemical Identifiers
Molecular Formula NaNO2
Molecular Weight (g/mol) 69
MDL Number MFCD00011118
Specifications
Boiling Point 320°C (1013 hPa)
Melting Point 284°C
Quantity 1 kg
Assay Percent Range min. 97.0%
Density 2.168 g/cm³ (20°C)
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H301-Toxic if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.