Learn More
SLC5A6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26407-1-AP-20UL
This item is not returnable.
View return policy
Description
Transports pantothenate, biotin and lipoate in the presence of sodium.Specifications
SLC5A6 | |
Polyclonal | |
Unconjugated | |
SLC5A6 | |
SLC5A6, SMVT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170551, 330064, 8884 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70247, Q5U4D8, Q9Y289 | |
Slc5a6 | |
SLC5A6 Fusion Protein Ag23907 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |