Learn More
SLC5A6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26407-1-AP-150UL
 View more versions of this product
Product Code. 16854875
  • 4652.90 SEK / 150µL
Description
Transports pantothenate, biotin and lipoate in the presence of sodium.
Specifications
Specifications
SLC5A6
Polyclonal
Unconjugated
SLC5A6
SLC5A6, SMVT
Rabbit
Antigen Affinity Chromatography
RUO
170551, 330064, 8884
-20°C
Liquid
Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70247, Q5U4D8, Q9Y289
Slc5a6
SLC5A6 Fusion Protein Ag23907
150 μL
Primary
Human, Mouse, Rat