Learn More
Thermo Scientific™ SL40F TX-1000 and F13-14X50C Centrifuge Package
Excellent ergonomics and high performance unit for daily sample preparation.
Brand:  Thermo Scientific™ 50146571
Product Code. 15314376
Includes: Thermo Scientific SL40F, 230V (Mfr PN: 75004542); TX-1000 rotor, Slide coat (Mfr PN: 75003017); TX-1000 buckets (set of 4) (Mfr PN: 75003001); TX-1000 ClickSeal biocontainment lids (set of 4) (Mfr PN: 75007309); Adapter, 50mL Conical (Mfr PN: 75003674); Adapter, 24 x 15mL, BIOFlex HC (Mfr PN: 75007306); F13S - 14x50c Carbon Fiber rotor (Mfr PN: 75003661)
This item is not returnable. View return policy
Description
Description
Revolutionary design provides reliability, simplicity and convenience when performing routine tasks.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products