Learn More
BRAND™ SILBERBRAND Automatic Burette with Lateral Safety Stopcock
Shop All Brand Products
Click to view available options
Includes:
2000 mL bottle
No bottle
Unit Size:
Each
Description
- Calibrated to deliver (TD, Ex)
- With needle-valve stopcock with PTFE spindle
- Schellbach stripe
- Automatic zeroing
Specifications
Specifications
Capacity (Metric) | 10 mL |
Graduations | 0.02 mL |
Includes | No bottle |
Stopcock | Lateral safety stopcock |
Warranty | 12 months |