Learn More
SCO2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21223-1-AP-150UL
This item is not returnable.
View return policy
Description
Synthesis of cytochrome c oxidase 2 was initially identified in yeast as one of two cytochrome c oxidase (COX) assembly proteins that enable the assembly of cytochrome c holoenzyme, a complex that catalyzes the transfer of reducing equivalents from cytochrome c to molecular oxygen and pumps protons across the inner mitochondrial membrane. Like their yeast homologs, the function of both SCO2 and SCO1 are dependent on copper ion binding. Recent studies suggest that SCO2 expression is regulated by p53, so that a decrease in p53 expression, such as in numerous tumors and cells lines, the drop in SCO2 expression leads to a shift from normal aerobic respiration towards the production of glycolytic ATP. Defects in the SCO2 protein are also associated with fatal infantile cardioencephalomyopathy and COX deficiency.Specifications
SCO2 | |
Polyclonal | |
Unconjugated | |
SCO2 | |
SCO1L, SCO2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100126824, 9997 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43819, Q8VCL2 | |
SCO2 | |
SCO2 Fusion Protein Ag15663 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |