missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCLT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14875-1-AP-150UL
This item is not returnable.
View return policy
Description
Adapter protein that links SCN10A to clathrin. Regulates SCN10A channel activity, possibly by promoting channel internalization.Specifications
SCLT1 | |
Polyclonal | |
Unconjugated | |
SCLT1 | |
CAP1A, hCAP 1A, SAP1, SCLT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132320, 67161 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G5E861, Q96NL6 | |
Sclt1 | |
SCLT1 Fusion Protein Ag6646 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction