missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAR1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22291-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to SAR1A.
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.Specifications
SAR1A | |
Polyclonal | |
Unconjugated | |
SAR1A | |
COPII associated small GTPase, GTP binding protein SAR1a, masra2, SAR1, SAR1 homolog A (S. cerevisiae), SAR1A, Sara, SARA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20224, 361842, 56681 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P36536, Q9NR31 | |
Sar1a | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction