Learn More
Eppendorf™ Sandwich-Mode Spacer
Required for stacking rotors F-45-72-8 and F-45-48-11
Brand:  Eppendorf™ 5301316005
390.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10397083
  • 391.56 SEK / Each
Description
  • Rotor spacer
  • For use with rotors F-45-72-8 and F-45-48-11
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Spacer
Rotor F-45-72-8 and F-45-48-11
Required for stacking rotors F-45-72-8 and F-45-48-11