Learn More
Bollé Safety Rush+ Platinum™ Safety Glasses
Shop All Bollé Safety Products
Click to view available options
Lens Tint:
Clear
Smoke
Frame Color:
Black
Red, Black
White, Blue
Unit Size:
1 pair
Each
Pack of 10
Description
- Upper protection
- Non-slip temples
- Adjustable non-slip bridge
- Safety glasses with clear lenses are essential indoors
Specifications
Specifications
Certifications/Compliance | CE European standards certificate available. |
Lens Type | Platimun™ |
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Glasses |
Frame Color | Red, Black |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | Yes |