Learn More
Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene PE Foam/PTFE Cap
Ideal general purpose bottle. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene PE Foam/PTFE Cap used for pharmaceutical, chemical, environmental, and food packaging and storing applications.
367.00 SEK - 810.00 SEK
Specifications
Color | Clear |
---|---|
Material | Glass |
Closure Color | White |
Liner Material | PE Foam/PTFE |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Closure Size | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Quantity | Price | Quantity & Availability | |||
11718749
|
Fisherbrand™
02911768 |
118 mL | 48-400 mm | 24/Cs. | |||||
11704329
|
Fisherbrand™
02911770 |
237 mL | 58-400 mm | 24/Cs. | |||||
11714329
|
Fisherbrand™
02911767 |
473 mL | 63-400 mm | 24/Cs. | |||||
11724329
|
Fisherbrand™
02911913 |
946 mL | 70-400 mm | 12/Cs. | |||||
11734329
|
Fisherbrand™
02911769 |
1893 mL | 83-400 mm | 6/Cs. | |||||
11553672
|
Fisherbrand™
02911766 |
3785 mL | 89-400 mm | 4/Cs. | |||||
Description
Clear / Flint Standard Wide Mouth Bottles feature a wide opening making them the ideal general purpose bottle. Applications include pharmaceutical, chemical, environmental, and food packaging and storing.
- Features wide mouth for easy transferring of contents
- Available in capacities ranging from 120 to 3840mL
Specifications
Clear | |
White | |
Yes | |
Round | |
PTFE/Polyethylene | |
Screw Cap |
Glass | |
PE Foam/PTFE | |
Wide | |
Standard Wide Mouth Bottle | |
Polypropylene |