Learn More
BUCHI FlashPure ID C18-WP Flash Chromatography Cartridges
FlashPure ID cartridges are designed for BUCHI Pure chromatography systems. Integrated RFID technology automatically transfers cartridge settings and data to the flash system.
76.00€ - 191.00€
Specifications
Material | Silica - C-18 WP |
---|---|
Type | Reveleris |
Includes | 1 Pc. |
For Use With (Application) | Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications |
Max. Pressure | 200 psig |
Includes: 1 Piece
Product Code | Brand | Bed Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight (Metric) | Price | Quantity & Availability | |||||
16390296
|
BUCHI
145169482 |
4 g | N/A | ||||||
16300306
|
BUCHI
145169483 |
12 g |
138.00€
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
16310306
|
BUCHI
145169484 |
40 g |
191.00€
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
|||||
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Silica - C-18 WP | |
1 Pc. | |
200 psig |
Reveleris | |
Ideal for Peptides, Proteins and Lipids and Other Large Molecule Purifications |