Learn More
Thermo Scientific™ Remel™ Gram Iodine
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
428.00 SEK - 1870.00 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11983322
|
Thermo Scientific™
R40077 |
Each | N/A | ||||||
11933972
|
Thermo Scientific™
R40056 |
Each |
428.00 SEK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12687589
|
Thermo Scientific™
R40057 |
5/Pk. | N/A | ||||||
Description
Thermo Scientific™ Remel Gram Iodine is a mordant used in Gram stain procedures.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
Specifications
Gram Iodine | |
CE Marked |