missing translation for 'onlineSavingsMsg'
Learn More

RB1CC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen RB1CC1
Concentration 0.38 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16810885
View Documents
Proteintech
21269-1-AP-20UL
20 μL
145.00€
20µL
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16800885
View Documents
Proteintech
21269-1-AP-150UL
150 μL
410.00€
150µL
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human RB1CC1.

The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

RB1CC1
Western Blot
Unconjugated
Rabbit
Human, Rat, Mouse
Q8TDY2, Q9ESK9
12421, 312927, 9821
RB1CC1 Fusion Protein Ag15739
Primary
-20°C
RB1CC1
0.38 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC
RB1CC1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.