Learn More
RALYL Mouse anti-Human, Unconjugated, Clone: 1F3F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67534-1-IG-150UL
Description
The RALYL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RALYL. This antibody recognizes human, mouse, rat, pig antigen. The RALYL antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
RALYL | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunofluorescence, ELISA | |
1F3F4 | |
Unconjugated | |
150 μL | |
138046 | |
IgG1 |