Learn More
Pyrex™ Quickfit™ Vacuum Receiver Adapter
Requires screwthread connector for operation. Pyrex™ Quickfit™ Vacuum Receiver Adapter can be used with Liebig condensers and other apparatuses with 24/29 cone size.
Brand:  Pyrex™ RA13/33/SC
56.81 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 11322095
  • 59.80€ / Each
Estimated Shipment: 20-06-2025
Explore more special offers
Description
Requires screwthread connector for operation. Pyrex™ Quickfit™ Vacuum Receiver Adapter can be used with Liebig condensers and other apparatuses with 24/29 cone size.
  • Delivery stem within cone
  • With sidearm for connection to a vacuum supply
  • Easy connection to flexible tubing via detachable plastic screwthread connector
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24/29
Borosilicate Glass
1Each
140mm
Glass joint adapter