Learn More
Quickfit™ Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere.
1980.00 SEK - 9785.00 SEK
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15768017
|
Quickfit
GB1/24 |
Reaction Vessel Bubbler | N/A | ||||||
15778017
|
Quickfit
GB1/29 |
Reaction Vessel Bubbler |
1980.00 SEK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
15537910
|
Quickfit
GB1/19 |
Reaction Vessel Bubbler |
9785.00 SEK
Pack of 5 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
|||||
Description
- Side arm to facilitate filling with mineral oil
- Bubblers have capacity of approx 5ml
Specifications
Reaction Vessel Bubbler |