Learn More
Cole-Parmer™ PTFE Manifold, 2 outlets
Organize your fluid paths with these thermoplastic manifolds
Brand:  Cole-Parmer™ 31521-03
 View more versions of this product
Product Code. 11795258
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • PTFE Manifold.
  • Chemically-inert PTFE handles the most aggressive chemical solutions.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Specifications
PTFE
6.03 cm
-129°C
1/8 in. NPT(F)
1.25 in.
1.25 in.
300°F
PTFE Manifold, 2 Outlets
2 3/8 in.
-200°F
3/8 in. NPT(F)
3.18 cm
3.18 cm
120 psi
150°C