Learn More
Jenway™ PTFE coated ATC probe for use with corrosive/aggressive samples
Automatic Temperature Compensation (ATC) probe used with Jenway pH Meters.
Brand:  Jenway™ 16158-30
1393.51 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11400579
  • 1480.00 SEK / Each
Only null left
Estimated Shipment: 29-08-2025
Description
  • For use with Models 3510, 3520 and 3540 pH Meters
  • Automataic Temperature Compensation probe for use with corrosive and aggressive samples
Specifications
Specifications
For use with aggressive/corrosive samples
PTFE-coated