Learn More
Fisherbrand™ PTFE Bottle Pourer
Fisherbrand™ PTFE Bottle Pourer permits direct pouring of all liquids from reagent bottles in a safe and consistent manner.
Brand: Fisherbrand™ 028.030
Description
- Totally inert with PTFE encapsulated viton neck seal
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
Fisherbrand™ PTFE Bottle Pourer | |
GL30 | |
Reagent Bottles |
PTFE | |
1/Ea. |