missing translation for 'onlineSavingsMsg'
Learn More

Protein C inhibitor Mouse anti-Human, Clone: 2H6G2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66030-1-IG-20UL

 View more versions of this product

Product Code. 16877535

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Protein C inhibitor
Monoclonal
1.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05154
SERPINA5
Protein C inhibitor Fusion Protein Ag18094
20 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
2H6G2
Unconjugated
SERPINA5
PAI3, PCI, PLANH3, PROCI, SERPINA5
Mouse
Protein A
RUO
5104
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.