Learn More
Prohibitin 2 Mouse anti-Human, CoraLite 488, Clone: 1D9C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66424-100UL
Description
The Prohibitin 2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Prohibitin 2. This antibody recognizes human, mouse, rat antigen. The Prohibitin 2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Prohibitin 2 | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
11331 | |
IgG1 |
Immunofluorescence | |
1D9C7 | |
Mouse | |
Primary | |
Human, Rat, Mouse |