Learn More
PRLR Mouse anti-Human, Unconjugated, Clone: 2E10G5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | PRLR |
---|---|
Clone | 2E10G5 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16869835
|
Proteintech
67292-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16859835
|
Proteintech
67292-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PRLR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRLR. This antibody recognizes human, mouse antigen. The PRLR antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
PRLR | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
5618 | |
Primary |
2E10G5 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human, Mouse | |
IgG1 |