Learn More
Memmert™ Potential-Free Contact (24 V / 2 A) with Socket to NAMUR NE 28 for Combined Error Message for UF55
Potential-free contact for combination error message (e.g. supply failure, sensor fault, fuse).
Brand: Memmert™ UF55H6
287.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- For combination error message
Specifications
UF55 Oven | |
Socket to NAMUR NE 28 | |
2 A |
Potential-free Contact | |
24 V |