Learn More
Potassium Thiocyanate, 0.1M, Chem Lab™
Product Code. 10293902
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Quantity 1 L
Concentration or Composition (by Analyte or Components) 0.1 mol/L
Density 1,01g/mL
compliance-icons
Product Identifier
  • Potassium thiocyanate 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : KCNS/H2O