Learn More
Thermo Scientific Chemicals Potassium permanganate, 98%, extra pure
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
Color | Black to Brown |
Physical Form | Lustrous Crystalline Powder |
Quantity | 5 kg |
Assay Percent Range | 98% |
Linear Formula | KMnO4 |
Fieser | 01,942; 02,348; 04,412; 05,562; 08,416; 09,388; 10,330; 11,440; 12,413; 13,258; 14,267; 15,273; 16,283 |
Merck Index | 15, 7776 |
SMILES | [K+].[O-][Mn](=O)(=O)=O |
IUPAC Name | potassium manganesoylolate |
Molecular Weight (g/mol) | 158.03 |