Learn More
Potassium Fluoride Dihydrate, 98%, Reagent grade, Honeywell™
Product Code. 15662160 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Reagent Grade, 98%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]
Specifications
Quantity 500 g
Linear Formula KF · 2H2O
UN Number UN1812
IUPAC Name potassium dihydrate fluoride
Formula Weight 94.13g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Potassium Fluoride Dihydrate