Learn More
VITLAB™ Polypropylene Measuring Scoop
Use this scoop for measuring and/or weighing. The Vitlab™ Polypropylene Measure Scoop is sold individually in one color or as a multi-color set.
Brand:  VITLAB™ 3961111
Product Code. 15312984
  • 176.00 SEK / Each
Only null left
Estimated Shipment: 11-08-2025
Description
  • Various colors
  • Also suitable as weighing scoops
  • Precisely formed filling edge and comfortable, stable handle
  • Easily readable volume specification on the upper side of the handle
Specifications
Specifications
Measuring Scoop
Polypropylene
100 mL
White, Red, Grey, Black, Yellow, Blue, Green, Bright Blue