Learn More
Buerkle™ Rapid Couplings, NW 6.4 mm, Male Connections, Angled Nipples with Hose Nozzle
Fast and safe connection for liquids, gases, compressed air and vacuum. Buerkle™ Rapid Couplings are ideal when connecting and separating hoses, machines, apparatus, containers and more.
Brand:  Buerkle™ 8775-1160
Product Code. 10430756
  • 313.36 SEK / Each
Only null left
In Stock
Description
  • Manufactured from polyoxymethylene
  • Maximum flow possible
  • Easy single-handed operation
  • Secure O-ring guarantees that couplings are leakproof
  • Operating pressure at 20°C: with vacuum up to 10 bar
  • Operating temperature from −40° to +80°C
  • Couplings simply connect to each other, automatically clicking into place and opening the valves
  • When disconnecting, the valves are automatically sealed, no leakage occurs
Specifications
Specifications
With valve
POM
9.5 mm Hose
1
Rapid coupling
80°C
9.5 mm
31 mm
-40°C
6.4 mm
10 bar
Rapid Coupling