Learn More
Buerkle™ Polychlorobutadiene Rubber O-Ring
Buerkle™ Polychlorobutadiene Rubber O-Ring for use with Buerkle™ Desiccators.
Brand: Buerkle™ 9791-0252
This item is not returnable.
View return policy
Description
- O-Ring
- Made of Polychlorobutadiene-rubber
Specifications
O-ring | |
For 250 mm I.D. Desiccators | |
Chloroprene Rubber | |
250mm I.D. desiccators |