Learn More
Nickel Electro™ Polycarbonate Hinged Gable Lid
Maximize bath area with a hinged lid that drips condensation back into bath. Nickel Electro™ Polycarbonate Hinged Gable Lid are transparent to allows samples to be viewed.
1315.00 SEK - 1395.00 SEK
Specifications
For Use With (Application) | Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy |
---|---|
Material | Polycarbonate |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11786577
|
Nickel Electro™
GL1-22 |
22 and 28 L Unstirred and Shaking Water Baths |
1395.00 SEK
Each |
Estimated Shipment: 18-08-2025
Log in to see stock. |
|||||
12758888
|
Nickel Electro™
GL1-8 |
8 and 10 L Unstirred and Shaking Water Baths |
1315.00 SEK
Each |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||||
Description
- Lightweight
- Semi-transparent for viewing samples
- Maximize bath area
Specifications
Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy | |
Polycarbonate |