Learn More
Memmert™ Plug-In Tube Extension for UF55, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ UF55B29718
522.03 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17599425
  • 549.00 SEK / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
UF55 Oven
Plug-in Tube Extension
O.D. 60.3 mm, I.D. 57 mm