Learn More
PLOD3 Mouse anti-Human, CoraLite 594, Clone: 2G12B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60058-100UL
Product Code. 16810876
  • 5106.85 SEK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The PLOD3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLOD3. This antibody recognizes human antigen. The PLOD3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
PLOD3
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence
2G12B8
CoraLite 594
100 μL
8985
IgG2a