Learn More
Platinum Iridium wire, 1.0mm (0.04in) dia, Thermo Scientific Chemicals
7850.00 SEK - 110320.00 SEK
Specifications
Chemical Name or Material | Platinum Iridium wire |
---|---|
Name Note | 1.0mm (0.04 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
MDL Number | MFCD02091683 |
Odor | Odorless |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11395407
|
Thermo Scientific Alfa Aesar
042678.BQ |
5 cm |
7850.00 SEK
5cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11385407
|
Thermo Scientific Alfa Aesar
042678.BU |
25 cm |
34460.00 SEK
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11305417
|
Thermo Scientific Alfa Aesar
042678.BY |
100 cm |
110320.00 SEK
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium wire | |
Pt:Ir; 90:10 wt% | |
Odorless | |
Wire |
1.0mm (0.04 in.) dia. | |
MFCD02091683 | |
≈0.85g/5cm |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures